Learn More
Enzo Life Sciences CD18 (human) monoclonal antibody (MEM-148) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX805043C100
Encompass_Preferred
Clone: MEM-148. Isotype: IgG1. Immunogen: Human peripheral blood mononuclear cells. Species reactivity: Human. Specificity: Recognizes free CD18 (β2 integrin) not associated with CD11. Applications: Flow Cytometry, IP, WB, FUNC. Purity: Protein A-affinity purified. Formulation: Liquid. In PBS, pH 7.4, containing 15 mM sodium azide.
Catalog No. 50-201-0041