Learn More
Biocare Medical, LLC CD19 Antibody - Predilute, 6.0 ml
SDP
Supplier:  Biocare Medical, LLC PM310AA
Encompass_Preferred
CD19 recognizes a 95kD cell surface glycoprotein, which is expressed by cells of the B-cell lineage and follicular dendritic cells. CD19 is a coreceptor of CD21 and is an important signal transduction molecule which is involved in the regulation of B lymphocyte development, activation and differentiation. Among the B-cell genes that are present in early B-cell development and are upregulated by Pax-5 are CD19 and CD79a. CD19 is found in pre-B cells, B cells (considered a pan Bcell antigen); first B-cell antigen after HLA-DR and follicular dendritic cells. CD19 is absent in plasma cells, most T-cell lymphomas and in lymphocyte predominant Hodgkin. It has been observed in lymphomas and leukemias but often weak/negative in follicular lymphoma or diffuse large B-cell lymphoma.
Catalog No. 50-831-19