Learn More
Bio Rad Laboratories CD19 antibody | Bu12, Mouse anti Human 0.2mg
Catalog No. NC3233504
Encompass
Mouse anti Human CD19 antibody, clone Bu12 recognizes human CD19, a 95 kDa cell surface glycoprotein, which is expressed on cells of the B cell lineage and follicular dendritic cells but absent on plasma cells. CD19 is an important signal transduction molecule which is involved in the regulation of B lymphocyte development, activation and differentiation.
Catalog No. NC3233504 Supplier Bio Rad Laboratories Supplier No. MCA2495
Mouse anti Human CD19 antibody, clone Bu12 recognizes human CD19, a 95 kDa cell surface glycoprotein, which is expressed on cells of the B cell lineage and follicular dendritic cells but absent on plasma cells. CD19 is an important signal transduction molecule which is involved in the regulation of B lymphocyte development, activation and differentiation.