Learn More
CAPRICO BIOTECHNOLOGIES INC CD19 iFluor 700
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4029175
Encompass_Preferred
The Invitrogen CD19 iFlor 700 is a mouse monoclonal antibody targeting human CD19, suitable for flow cytometry. It binds native and recombinant proteins, produced from NALM-6 cells, and comes in 100 μL of 0.2 mg/mL for 100 tests. Store at 4°C, avoiding light.
Catalog No. 50-227-9045