Learn More
CAPRICO BIOTECHNOLOGIES INC CD19 iFluor 700 Antibody, Clone SJ25C1, 100 Tests
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4029195
Encompass_Preferred
The CD19 iFluor 700 Antibody from Caprico Biotechnologies is a mouse monoclonal antibody that targets human CD19. It is conjugated to iFluor 700 and is suitable for flow cytometry applications. This antibody comes at a concentration of 0.5 mg/mL with 0.09% sodium azide as a preservative. Each vial contains 100 tests. The clone for this antibody is SJ25C1. Overall, this CD19 antibody allows detection of CD19 positive cells such as B cells via flow cytometry when used properly.
Catalog No. 50-227-9044