Learn More
CAPRICO BIOTECHNOLOGIES INC CD19 iFluor488 Antibody
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4029115
Encompass_Preferred
The Caprico CD19 iFluor 488 antibody is a mouse monoclonal antibody that specifically binds to human CD19. It is conjugated to iFluor 488, a green fluorescent dye, and is suitable for flow cytometry, immunofluorescence microscopy, and immunohistochemistry. This antibody recognizes the 95 kDa CD19 cell surface antigen found on B lymphocytes and follicular dendritic cells. It can be used for analyzing and isolating B cell populations. The antibody is available in multiple sizes ranging from 25 μg to 1 mg.
Catalog No. 50-227-9050