Learn More
BPS Bioscience Inc CD19 Knockout Raji Cell Line, 2 vials
SDP
Supplier:  BPS Bioscience Inc 82166
Encompass
CD19 Knockout Raji Cell Line is a Raji cell line where CD19 has been genetically removed from Raji cells using CRISPR/Cas9 genome editing. Use of this cell line is for research purposes only.
Catalog No. NC2869580
Add to Cart
Request a Quote