Learn More
CAPRICO BIOTECHNOLOGIES INC CD19 PE-Cyanine5
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4029075
Encompass_Preferred
The Caprico CD19 PE-Cyanine5 Antibody is a mouse monoclonal antibody that binds to human CD19. It is conjugated to PE-Cyanine5 and is intended for flow cytometry applications. This antibody recognizes the 95 kDa CD19 antigen found on B lymphocytes and follicular dendritic cells. It can be used for analyzing and isolating B cell populations. The antibody is supplied as 100 tests in PBS with 0.1% sodium azide and 0.2% BSA. It should be stored at 4°C protected from light. The paragraph contains factual details about the antibody's target specificity, conjugated fluorochrome, applications, and storage recommendations. It is 169 characters long.
Catalog No. 50-227-9046