Learn More
Sino Biological Recombinant Human CD19 Protein (ECD, His Tag)
Supplier:  Sino Biological 11880H08H500
Encompass_Preferred
A DNA sequence encoding the human CD19 (P15391-1) extracellular domain (Met1-Lys291) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-6447