Learn More
Sino Biological Recombinant Mouse CD19 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 50510M08HB100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse CD19 (NP_033974.2) (Met1-Gly287) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6257