Learn More
Enzo Life Sciences CD19 Recombinant monoclonal antibody (4G7)(Rabbit IgG kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS4120200
Encompass_Preferred
Clone: 4G7. Isotype: IgGκ. Immunogen: Human CCL (chronic lymphocytic leukemia) cells. Species reactivity: Human. Applications: Flow Cytometry, IF. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-2939