Learn More
Enzo Life Sciences CD19 Recombinant monoclonal antibody (FMC63) (Mouse IgG2a kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS4150200
Encompass_Preferred
Clone: FMC63. Isotype: IgG2aκ. Immunogen: Human prolymphocytic leukaemia cell line JVM3. Species reactivity: Human. Applications: Flow Cytometry, IF. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-2942