Learn More
Enzo Life Sciences CD19 Recombinant monoclonal antibody (HD37) (Rabbit IgG kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS4160200
Encompass_Preferred
Clone: HD37. Isotype: IgGκ. Immunogen: Cells from a hairy cell leukemia patient. Species reactivity: Human. Applications: Flow Cytometry, IF, IHC, IP, FUNC. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-2943