Learn More
Cayman Chemical CD19CTerm RabbIt Monclon 100uL
SDP
Catalog No. 502948351
Encompass_Preferred
A RIPK2 inhibitor inhibits RIPK2 by 65 in a kinase assay at 10 UM inhibits VEGFR3 by greater than 90 as well as 14 additional kinases by 60-89 in a panel of 344 kinases at 10 UM reduces colonic damage in a mouse model of TNBS-induced colitis at 7.5 and 145 mg/kg twice per day a prodrug
Catalog No. 50-294-8351 Supplier Cayman Chemical Supplier No. 32269100 UL
A RIPK2 inhibitor inhibits RIPK2 by 65 in a kinase assay at 10 UM inhibits VEGFR3 by greater than 90 as well as 14 additional kinases by 60-89 in a panel of 344 kinases at 10 UM reduces colonic damage in a mouse model of TNBS-induced colitis at 7.5 and 145 mg/kg twice per day a prodrug