Learn More
ACROBiosystems CD19 Protein, Cynomolgus and Rhesus Macaque, Fc Tag, 1 mg
Catalog No. 501658601
Encompass_Preferred
This B-lymphocyte antigen CD19 protein is expressed in human HEK293 cells and contains amino acids Pro 20-Lys 292 of the cynomolgus and rhesus macaque CD19 sequence. It is designed with a human IgG1 Fc tag at the C-terminus and is provided in a lyophilized powder format.
  • Purity ≥95% as determined by SDS-PAGE
  • Endotoxin levels <0.01 EU per μg
  • Calculated molecular weight of 56.7 kDa, migrating as 65-95 kDa due to glycosylation
  • Formulated in Tris with glycine, arginine, NaCl, and trehalose at pH 7.5
  • Stable for 12 months at -20°C to -70°C in lyophilized state
  • Validated for use in signal transduction and receptor complex studies
Catalog No. 50-165-8601 Supplier ACROBiosystems Supplier No. CD9C52511MG
This B-lymphocyte antigen CD19 protein is expressed in human HEK293 cells and contains amino acids Pro 20-Lys 292 of the cynomolgus and rhesus macaque CD19 sequence. It is designed with a human IgG1 Fc tag at the C-terminus and is provided in a lyophilized powder format.
  • Purity ≥95% as determined by SDS-PAGE
  • Endotoxin levels <0.01 EU per μg
  • Calculated molecular weight of 56.7 kDa, migrating as 65-95 kDa due to glycosylation
  • Formulated in Tris with glycine, arginine, NaCl, and trehalose at pH 7.5
  • Stable for 12 months at -20°C to -70°C in lyophilized state
  • Validated for use in signal transduction and receptor complex studies