Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 909RBM6P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: C1A/1506R. Clonality: Monoclonal. Host: Rabbit. Isotype: IgG kappa. Positive Control: MOLT-4 cells. Paracortex in a tonsil or a reactive lymph node. Cellular Staining: Cell surface. Buffer: 10mM PBS with or without 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human full-length CD1a protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 49kDa.Chromosome: 1q23.1. Gene Symbol: CD1A. Gene ID: 909. Ensemble: ENSG00000158477. Uniprot: NP_001754.
Catalog No. 50-188-5320