Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 909MSM3P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: O10 + C1A/711. Clonality: Monoclonal. Host: Mouse. Isotype: IgG. Positive Control: MOLT-4 cells. Paracortex in a tonsil or a reactive lymph node. Cellular Staining: Cell surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Human thymus cells (O10); Recombinant human CD1a protein (C1A/711). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 49kDa.Chromosome: 1q23.1. Gene Symbol: CD1A. Gene ID: 909. Ensemble: ENSG00000158477. Uniprot: NP_001754.
Catalog No. 50-188-5319