Learn More
Aviva Systems Biology Corp CD1A Antibody - C-terminal region (ARP63049_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63049_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CD1A. It was validated on Western Blot. . Alternate names: CD1, FCB6, HTA1, R4, T6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Sheep
Catalog No. 50-287-854