Learn More
CAPRICO BIOTECHNOLOGIES INC CD1a PerCP-Cyanine5.5
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4115065
Encompass_Preferred
The CD1a (PerCP-Cyanine5.5) antibody from is a mouse monoclonal antibody that specifically binds to human CD1a. CD1a is a transmembrane glycoprotein expressed on cortical thymocytes, dendritic cells, and Langerhans cells. This antibody is conjugated to PerCP-Cyanine5.5 and is recommended for flow cytometry applications such as analyzing dendritic cell populations. It is supplied as 100 tests in PBS with 0.1% sodium azide and 0.2% BSA. The concentration is 0.2 mg/mL.
Catalog No. 50-227-8971