Learn More
Aviva Systems Biology Corp CD1B Antibody - N-terminal region (ARP63421_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63421_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CD1B. It was validated on Western Blot. . Alternate names: CD1, CD1A, MGC125990, MGC125991, R1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Sheep
Catalog No. 50-287-964