Learn More
Cayman Chemical CD1d MoncLNal ANTIBDYClo 100ug
SDP
Catalog No. 502911114
Encompass_Preferred
Immunogen: Recombinant human CD1d extracellular domain/β2-microglobulin complex • Host: Mouse • Species Reactivity: (+) Human • Cross Reactivity: (+) CD1d • Applications: ELISA, FC, WB • MW = ~52 kDa
Catalog No. 50-291-1114 Supplier Cayman Chemical Supplier No. 34936100 UG
Immunogen: Recombinant human CD1d extracellular domain/β2-microglobulin complex • Host: Mouse • Species Reactivity: (+) Human • Cross Reactivity: (+) CD1d • Applications: ELISA, FC, WB • MW = ~52 kDa