Learn More
ABclonal Technology CD1d Rabbit mAb
SDP
Catalog No. 5000012946
Encompass_Preferred
Enables T cell receptor binding activity and endogenous lipid antigen binding activity Involved in NK T cell differentiation and regulation of immature T cell proliferation in thymus Acts upstream of or within several processes including antigen processing and presentation exogenous lipid antigen via MHC class Ib positive regulation of cytokine production and positive regulation of leukocyte activation Located in endosome external side of plasma membrane and lysosome Is expressed in forebrain intestine jaw bone and liver Human ortholog(s) of this gene implicated in pulmonary tuberculosis Orthologous to human CD1D (CD1d molecule)
Catalog No. 50-000-12946 Supplier ABclonal Technology Supplier No. A26678100UL
Enables T cell receptor binding activity and endogenous lipid antigen binding activity Involved in NK T cell differentiation and regulation of immature T cell proliferation in thymus Acts upstream of or within several processes including antigen processing and presentation exogenous lipid antigen via MHC class Ib positive regulation of cytokine production and positive regulation of leukocyte activation Located in endosome external side of plasma membrane and lysosome Is expressed in forebrain intestine jaw bone and liver Human ortholog(s) of this gene implicated in pulmonary tuberculosis Orthologous to human CD1D (CD1d molecule)