Learn More
Aviva Systems Biology Corp CD2 Antibody - C-terminal region (ARP62984_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62984_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CD2. It was validated on Western Blot. . Alternate names: FLJ46032, SRBC, T11ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-287-828