Learn More
CAPRICO BIOTECHNOLOGIES INC CD2 Antibody, 4002045, APC-conjugated, Clone S5.2
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4002045
Encompass_Preferred
The Caprico CD2 Antibody 4002045 APC Conjugated is a mouse monoclonal antibody that specifically binds to human CD2. It is conjugated to APC fluorochrome and is suitable for flow cytometry, immunofluorescence microscopy and immunohistochemistry assays. This antibody recognizes the T11TS CD2 epitope and has been tested for cross-reactivity in rhesus, cynomolgus and baboon samples. It is provided at a concentration of 0.2 mg/mL in PBS with 0.1% sodium azide and 2 mg/mL BSA. The clone is S5.2.
Catalog No. 50-227-9051