Learn More
CAPRICO BIOTECHNOLOGIES INC CD2 Antibody, APC-Cyanine7, 4002095
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4002095
Encompass_Preferred
The Caprico CD2 Antibody APC-Cyanine7 is a monoclonal antibody that specifically binds to human CD2. It is conjugated to APC-Cyanine7 and is suitable for flow cytometry, immunohistochemistry, and immunofluorescence. This antibody recognizes the T11TS CD2 epitope and has been tested for cross-reactivity in rhesus, cynomolgus, and baboon samples. It is available in 25 tests, 100 tests, and bulk sizes. The recommended usage is 5 μL per test. This antibody can be used for research purposes to identify T cells and NK cells.
Catalog No. 50-227-9055