Learn More
CAPRICO BIOTECHNOLOGIES INC CD2 PE-Cyanine7, 4002085
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4002085
Encompass_Preferred
The Caprico CD2 PE-Cyanine7 antibody is a mouse monoclonal antibody that recognizes human CD2. It is conjugated to PE-Cyanine7 and is suitable for flow cytometry applications. This antibody recognizes the T11 target on human T cells and NK cells. It can be used to identify T cell populations and subsets. The antibody is supplied as 100 tests in PBS with 0.1% sodium azide and 0.2% BSA. Each test contains 100 μL of antibody. The product should be stored at 4°C protected from light. This antibody is for research use only.
Catalog No. 50-227-9052