Learn More
Sino Biological Recombinant Human CD2 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10982H08H100
Encompass_Preferred
A DNA sequence encoding the human CD2 (NP_001758.2) extracellular domain (Met 1-Asp 209) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-6288