Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 931MSM3XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: SPM618. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: Daudi, Raji,, U266,, human lymphocytes. Lymph nodes, tonsils. Cellular Staining: Predominantly cell surface with some cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human MS4A1 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 33-37kDa.Chromosome: 11q12.2. Gene Symbol: MS4A1. Gene ID: 931. Ensemble: ENSG00000156738. Uniprot: NP_690605.
Catalog No. 50-188-5333