Learn More
ACROBiosystems ACROBiosystems Biotinylated Human CD20 / MS4A1 Full Length Protein, His,Avitag™ (HEK293)
Supplier:  ACROBiosystems CD0H82E520UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human CD20 Full Length, His,Avitag (HEK293) (CD0-H82E5) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Pro 297 (Accession # P11836-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 36.9 kDa.
Catalog No. 50-195-6414