Learn More
Biogenex Laboratories CD20 : L-26 : 6mL
SDP
Supplier:  Biogenex Laboratories AM2385M
Encompass_Preferred
This antibody reacts with a formalin-resistant intracytoplasmic epitope found in the majority of B cells which is now considered to be the CD20 antigen, a pan-B cell marker. The antibody primarily recognizes a 33 kD polypeptide B cell component and also a minor 30 kD cellular antigen. The staining pattern is consistent with pan-B reactivity, producing staining for B cells in lymphoid and peripheral blood tissue. This antibody intensely stains germinal centers and B immunoblasts in lymphoid tissue.The L26 antibody is widely regarded as the most selective B cell marker currently available. L26 has shown no cross-reactivity with nonlymphoid tissues. L26 may prove to be a useful marker for L&H variants of Reed-Sternberg cells of Hodgkin's lymphomas where reactive pattern is distinct from other Reed-Sternberg variants.
Catalog No. 50-197-8755