Learn More
Enzo Life Sciences CD20 Recombinant monoclonal antibody (IGEL/1497R) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS3450100
Encompass_Preferred
Clone: IGEL/1497R. Isotype: IgGκ. Immunogen: Recombinant human full-length CD20. Species reactivity: Human. Applications: Flow Cytometry, IF, IHC (PS). Purity: Protein A affinity purified. Formulation: Liquid. In 1X PBS containing 0.1 mg/ml BSA (US sourced) and 0.05% sodium azide.
Catalog No. 50-201-2875