Learn More
Medchemexpress LLC CD20/MS4A1 Mouse P 50ug
SDP
Catalog No. 5000196091
Encompass_Preferred
Recombinant mouse CD20 (MS4A1) protein expressed in P. pastoris and provided with an N-terminal 6xHis tag. The protein is intended for research use in studies of B-cell signaling, antibody validation, binding assays, and biochemical characterization; it retains functional properties related to calcium influx regulation and membrane multimerization.
  • Expressed in P. pastoris for eukaryotic post-translational processing.
  • Contains an N-terminal 6xHis tag to facilitate purification and detection.
  • Available in multiple microgram sizes to suit different assay scales.
  • Suitable for biochemical, binding, and antibody validation assays.
  • Retains functional features involved in calcium influx regulation and multimer formation.
Catalog No. 50-001-96091 Supplier Medchemexpress LLC Supplier No. HYP7175850UG
Recombinant mouse CD20 (MS4A1) protein expressed in P. pastoris and provided with an N-terminal 6xHis tag. The protein is intended for research use in studies of B-cell signaling, antibody validation, binding assays, and biochemical characterization; it retains functional properties related to calcium influx regulation and membrane multimerization.
  • Expressed in P. pastoris for eukaryotic post-translational processing.
  • Contains an N-terminal 6xHis tag to facilitate purification and detection.
  • Available in multiple microgram sizes to suit different assay scales.
  • Suitable for biochemical, binding, and antibody validation assays.
  • Retains functional features involved in calcium influx regulation and multimer formation.