Learn More
Medchemexpress LLC CD20/MS4A1 Mouse P 5ug | 5ug
SDP
Catalog No. 5000196214
Encompass_Preferred
CD20/MS4A1 protein regulates cellular calcium influx essential for B-lymphocyte development differentiation and activation It activates store-operated calcium channels allowing calcium influx through B-cell receptor/BCR activation CD20/MS4A1 protein forms homotetramers and interacts with cell surface IgM chains the antigen-binding parts of the BCR CD20/MS4A1 Protein Mouse (P pastoris His) is the recombinant mouse-derived CD20/MS4A1 protein expressed by P pastoris with N-6 His labeled tag
Catalog No. 50-001-96214 Supplier Medchemexpress LLC Supplier No. HYP717585UG
CD20/MS4A1 protein regulates cellular calcium influx essential for B-lymphocyte development differentiation and activation It activates store-operated calcium channels allowing calcium influx through B-cell receptor/BCR activation CD20/MS4A1 protein forms homotetramers and interacts with cell surface IgM chains the antigen-binding parts of the BCR CD20/MS4A1 Protein Mouse (P pastoris His) is the recombinant mouse-derived CD20/MS4A1 protein expressed by P pastoris with N-6 His labeled tag