Learn More
Enzo Life Sciences CD20 Recombinant monoclonal antibody (10F381) (Human IgG1 kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS4190200
Encompass_Preferred
Clone: 10F381 (Rituximab). Isotype: IgG1κ. Immunogen: Human lymphoblastoid cell line SB. Species reactivity: Human. Applications: Flow Cytometry. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-2946