Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio Rad Laboratories CD20-STARBRIGHT ULTRAVIOLET575

Catalog No. NC3585380
Encompass

  • Target antigen is CD20 (Human)
  • Clone 2H7
  • Host species is Mouse
  • Isotype is IgG2b
  • Conjugated to StarBright UltraViolet 575
  • Designed for flow cytometry applications
  • Reacts with Human and Rhesus Monkey CD20
  • Excitation maximum at 340 nm
  • Emission maximum at 569 nm
  • Product form is purified IgG, liquid
  • Preparation using affinity chromatography on Protein G
  • Buffer solution is phosphate buffered saline
  • Contains 0.09% sodium azide as preservative
  • Stabilizers include 1% Bovine Serum Albumin, 0.1% Pluronic F68, 0.1% PEG 3350, 0.05% Tween 20
  • Unit size is 100 tests/0.5 ml
  • Store at +4°C, do not freeze
  • Shipped at ambient temperature

Catalog No. NC3585380 Supplier Bio Rad Laboratories Supplier No. MCA1710SBUV575
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Target antigen is CD20 (Human)
  • Clone 2H7
  • Host species is Mouse
  • Isotype is IgG2b
  • Conjugated to StarBright UltraViolet 575
  • Designed for flow cytometry applications
  • Reacts with Human and Rhesus Monkey CD20
  • Excitation maximum at 340 nm
  • Emission maximum at 569 nm
  • Product form is purified IgG, liquid
  • Preparation using affinity chromatography on Protein G
  • Buffer solution is phosphate buffered saline
  • Contains 0.09% sodium azide as preservative
  • Stabilizers include 1% Bovine Serum Albumin, 0.1% Pluronic F68, 0.1% PEG 3350, 0.05% Tween 20
  • Unit size is 100 tests/0.5 ml
  • Store at +4°C, do not freeze
  • Shipped at ambient temperature

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.