Learn More
Genscript Corporation CD200 R1/CRTR2 hFc Chimera Hu
Catalog No. 5000057213
Encompass
CD200Fc a chimeric molecule including the extracellular domain of CD200 and a murine IgG2a Fc region regulates immune responses following engagement of a cell surface receptor CD200R expressed on cells of the myeloid and T cell lineage A recent report focused attention on a family of CD200Rs but concluded that only one member used CD200 as its ligand
Catalog No. 50-000-57213 Supplier Genscript Corporation Supplier No. Z04167100
CD200Fc a chimeric molecule including the extracellular domain of CD200 and a murine IgG2a Fc region regulates immune responses following engagement of a cell surface receptor CD200R expressed on cells of the myeloid and T cell lineage A recent report focused attention on a family of CD200Rs but concluded that only one member used CD200 as its ligand