Learn More
ACROBiosystems Recombinant Protein;Human CD200 / OX-2 Protein, Fc Tag;HEK293;100UG;OX2,CD200,MOX1,MOX2,MRC,OX-2,My033,OX2
Catalog No. 501658954
Encompass_Preferred
Recombinant Protein;Human CD200 / OX-2 Protein, Fc Tag;100UG;Human CD200, Fc Tag (OX2-H5251) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gly 232 (Accession # AAH22522).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 48.6 kDa. The protein migrates as 50-68 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD200, Fc Tag (Cat. No. OX2-H5251) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD200 R1, Fc Tag (Cat. No. CR2-H82F4) with a linear range of 0.5-7.8 ng/mL (QC tested).
Catalog No. 50-165-8954 Supplier ACROBiosystems Supplier No. OX2H5251100UG
Recombinant Protein;Human CD200 / OX-2 Protein, Fc Tag;100UG;Human CD200, Fc Tag (OX2-H5251) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gly 232 (Accession # AAH22522).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 48.6 kDa. The protein migrates as 50-68 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD200, Fc Tag (Cat. No. OX2-H5251) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD200 R1, Fc Tag (Cat. No. CR2-H82F4) with a linear range of 0.5-7.8 ng/mL (QC tested).