Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human CD200 / OX-2 Protein, Fc Tag, Avi Tag (Avitag™);HEK293;200UG;OX2,CD200,MOX1,MOX2,MRC,OX-2,My033,OX2
Supplier:  ACROBiosystems OX2H82F1200UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human CD200 / OX-2 Protein, Fc Tag, Avi Tag (Avitag™);200UG;Biotinylated Human CD200, Fc Tag, Avi Tag (OX2-H82F1) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gly 232 (Accession # P41217-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 51.4 kDa. The protein migrates as 65-76 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7979