Learn More
Enzo Life Sciences CD200R Recombinant monoclonal antibody (OX108) (Rabbit IgG kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS4200200
Encompass_Preferred
Clone: OX108. Isotype: IgGκ. Immunogen: Recombinant protein - hCD200RCD4d3+4. Species reactivity: Human. Applications: Flow Cytometry. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-2947