Learn More
Sino Biological CD200RLa / CD200R1L Antibody (Biotin), Mouse MAb 100ug
Supplier:  Sino Biological 11620MM09B100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD200RLa / CD200R1L (rh CD200RLa / CD200R1L; Catalog#11620-H08H; AAT00538.1; Met1-Leu239) and then biotinylated. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and then biotinylated.
Catalog No. 50-224-8273