Learn More
ACROBiosystems ACROBiosystems Human CD206 / MMR Protein, His Tag
Catalog No. 501956472
Encompass_Preferred
Recombinant Protein;100UG;Human CD206, His Tag (CD6-H52H9) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Ala 1389 (Accession # P22897-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 158.6 kDa. The protein migrates as 150-190 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6472 Supplier ACROBiosystems Supplier No. CD6H52H9100UG
Recombinant Protein;100UG;Human CD206, His Tag (CD6-H52H9) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Ala 1389 (Accession # P22897-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 158.6 kDa. The protein migrates as 150-190 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.