Learn More
Enzo Life Sciences Langerin (human) monoclonal antibody (DCGM4/122D5) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS2420100
Encompass_Preferred
Clone: DCGM4/122D5. Isotype: IgG1. Immunogen: In vitro-derived human DCs from CD34+ cells in GM-CSF+TNF-α. Species reactivity: Human Dog. Specificity: Recognizes an extracellular epitope in carbohydrate recognition domain of Langerin. Applications: Flow Cytometry, IHC (FS) Internalization. Purity: Purified using QMA Hyper D ion exchange chromatography. Formulation: Liquid. In TRIS-NaCl, pH 8.
Catalog No. 50-201-2789