Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 30835MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: C209/1781. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b kappa. Positive Control: THP-1 or HeLa Cells. Small Intestine, Uterus or Lymph Node. Cellular Staining: Secreted and Cell Surface. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human CD209 protein fragment. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 45kDa.Chromosome: 19p13. Gene Symbol: CD209. Gene ID: 30835. Ensemble: ENSG00000090659. Uniprot: NP_001138371.
Catalog No. 50-188-4768