Learn More
Aviva Systems Biology Corp CD209 Antibody - N-terminal region (ARP42586_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42586_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CD209. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human CD209. Alternate names: CDSIGN, CLEC4L, DC-SIGN, DC-SIGN1, MGC129965ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-285-009