Learn More
CAPRICO BIOTECHNOLOGIES INC CD21 FITC, 4097015
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4097015
Encompass_Preferred
The Caprico CD21 FITC antibody is a mouse monoclonal antibody that targets human CD21. It is conjugated to fluorescein isothiocyanate (FITC) and is suitable for flow cytometry, immunofluorescence microscopy and immunohistochemistry. This antibody recognizes the C3d receptor and can be used to identify B lymphocytes, follicular dendritic cells and some virus infected cells. It is available in 100 tests of 50μl purified antibody with 0.09% sodium azide and 0.2% gelatin.
Catalog No. 50-227-9065