Learn More
Medchemexpress LLC CD21 Human HEK293 10ug
SDP
Catalog No. 5000181835
Encompass_Preferred
CD21 Protein a receptor for complement C3 Epstein-Barr virus and HNRNPU activates B lymphocytes and aids Epstein-Barr virus entry in microbial infection Its engagement with these ligands highlights its vital role in immune responses and interactions regulating B-cell activation and host defense against infections CD21 Protein Human (HEK293 His) is the recombinant human-derived CD21 protein expressed by HEK293 with C-His C-10 His labeled tag
Catalog No. 50-001-81835 Supplier Medchemexpress LLC Supplier No. HYP7289010UG
CD21 Protein a receptor for complement C3 Epstein-Barr virus and HNRNPU activates B lymphocytes and aids Epstein-Barr virus entry in microbial infection Its engagement with these ligands highlights its vital role in immune responses and interactions regulating B-cell activation and host defense against infections CD21 Protein Human (HEK293 His) is the recombinant human-derived CD21 protein expressed by HEK293 with C-His C-10 His labeled tag