Learn More
Enzo Life Sciences CD21 monoclonal antibody (BB6-11C9.6) (Biotin) (500µg)
SDP
Supplier:  Enzo Life Sciences ENZABS8120500
Encompass_Preferred
Clone: BB6-11C9.6. Isotype: IgG1. Species reactivity: Porcine. Applications: Flow Cytometry. Purity: Purified. Formulation: Liquid. In PBS containing sodium azide.
Catalog No. 50-201-3336