Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 933MSM5P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: BLCAM/1795. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Raji, Daudi, IM9, JY25 and human peripheral blood lymphocytes. Human Tonsil. Cellular Staining: Cell Surface. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human BL-CAM/CD22 protein (aa52-178). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 130-140kDa.Chromosome: 19q13.1. Gene Symbol: CD22. Gene ID: 933. Ensemble: ENSG00000012124. Uniprot: NP_001172030.
Catalog No. 50-188-5336