Learn More
Aviva Systems Biology Corp CD22 Antibody - N-terminal region (ARP59863_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59863_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CD22. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human CD22. Alternate names: FLJ22814, MGC130020, SIGLEC-2, SIGLEC2ApplicationWB. Species reactivity: Human
Catalog No. 50-286-856