Learn More
Sino Biological Recombinant Anti-Siglec-2/CD22 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 51177R026100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD22 (rM CD22; Catalog#51177-M08H; BAE33829.1; Met1-Arg708).
Catalog No. 50-222-3000